CAS RN 82874-83-7
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Dimethyl (trityloxyimino)malonate, 98%, Thermo Scientific™
CAS: 82874-83-7 Synonym: (Trityloxyimino)malonic acid dimethyl ester; Dimethyl 2-[(triphenylmethoxy)imino]malonate