accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CCH3CH3

CAS RN 829-26-5

IUPAC Name: 2,3,6-trimethylnaphthalene
Synonyms: 2,3,6-trimethylnaphthalene
Molecular Weight (g/mol): 170.26
Molecular Formula: C13H14
InChi Key: UNBZRJCHIWTUHB-UHFFFAOYSA-N
SMILES: CC1=CC=C2C=C(C)C(C)=CC2=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results