accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3H3CN

CAS RN 83-24-9

IUPAC Name: 2,5-dimethyl-1-phenyl-1H-pyrrole
Synonyms: 2,5-dimethyl-1-phenylpyrrole
Molecular Weight (g/mol): 171.24
Molecular Formula: C12H13N
InChi Key: JNXIFVSGXLGULI-UHFFFAOYSA-N
SMILES: CC1=CC=C(C)N1C1=CC=CC=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results