Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,5-Dimethyl-1-phenylpyrrole 98.0+%, TCI America™
SDP

Catalog No. P16155G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P16155G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P16155G Supplier TCI America Supplier No. P16155G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 83-24-9
Molecular Formula C12H13N
Molecular Weight (g/mol) 171.243
InChI Key JNXIFVSGXLGULI-UHFFFAOYSA-N
Synonym 2,5-dimethyl-1-phenyl-1h-pyrrole, 1h-pyrrole, 2,5-dimethyl-1-phenyl, pyrrole, 2,5-dimethyl-1-phenyl, 1-phenyl-2,5-dimethylpyrrole, acmc-1bkez, pyrrole,5-dimethyl-1-phenyl, 1h-pyrrole,5-dimethyl-1-phenyl, 1-phenyl-2,5-dimethyl-1h-pyrrole, 2,5-dimethyl-1-phenyl-1h-pyrrole #, pyrrole, 2,5-dimethyl-1-phenyl-8ci
PubChem CID 66518
IUPAC Name 2,5-dimethyl-1-phenylpyrrole
SMILES CC1=CC=C(N1C2=CC=CC=C2)C

Specifications

Melting Point 49°C
Boiling Point 160°C
Quantity 5 g
Formula Weight 171.24
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2,5-Dimethyl-1-phenylpyrrole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.