accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 83-32-9

IUPAC Name: 1,2-dihydroacenaphthylene
Synonyms: acenaphthene naphthyleneethylene 1,8-dihydroacenaphthalene 1,8-ethylenenaphthalene ethylenenaphthalene peri-ethylenenaphthalene
Molecular Weight (g/mol): 154.21
Molecular Formula: C12H10
InChi Key: CWRYPZZKDGJXCA-UHFFFAOYSA-N
SMILES: C1CC2=C3C1=CC=CC3=CC=C2
Boiling Point 
  • (2)
  • (6)
  • (3)
Percent Purity 
  • (3)
  • (2)
  • (6)
Quantity 
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
Physical Form 
  • (3)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (1)
  • (1)
  • (3)
  • (1)
  • (3)
  • (5)
  • (2)
  • (6)
  • (3)
  • (3)
  • (2)
  • (6)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)