Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Acenaphthene, MilliporeSigma™ Supelco™

Catalog No. 111016701 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
5000 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-6701 5000 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-6701 Supplier MilliporeSigma Supelco Supplier No. 48500U
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Environmental
TRUSTED_SUSTAINABILITY

Specifications

Packaging Ampule
CAS 83-32-9
Vapor Pressure 10 mmHg (131°C)
Vapor Density 5.32 (vs. air)
Flash Point 125°C
Physical Form Neat
Linear Formula C12H10
MDL Number MFCD00003807
Melting Point 90°C to 94°C
Boiling Point 279°C
Synonym 1,8-Ethylenenaphthalene
Type Organic Pollutant Standard
Quantity 5000 mg
Formula Weight 154.21
Grade Analytical Standard
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.