CAS RN 83-39-6
IUPAC Name:
1H-imidazole-4,5-dicarboxamide
Synonyms:
1H-imidazole-4,5-dicarboxamide
Molecular Weight (g/mol):
154.13
Molecular Formula:
C5H6N4O2
InChi Key:
NNWAARLSYSBVPB-UHFFFAOYSA-N
SMILES:
NC(=O)C1=C(N=CN1)C(N)=O
1
–
1
of
1
results
1
Imidazole-4,5-dicarboxamide 97.0+%, TCI America™
CAS: 83-39-6 Molecular Formula: C5H6N4O2 Molecular Weight (g/mol): 154.13 MDL Number: MFCD00047022 InChI Key: NNWAARLSYSBVPB-UHFFFAOYSA-N Synonym: Glycalbylamide PubChem CID: 65745 IUPAC Name: 1H-imidazole-4,5-dicarboxamide SMILES: NC(=O)C1=C(N=CN1)C(N)=O