Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Imidazole-4,5-dicarboxamide 97.0+%, TCI America™
SDP

Numéro de catalogue. I043525G missing translation for 'brandsLinkText'
Modifier l'affichage
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
I043525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Numéro de catalogue. I043525G Fournisseur TCI America Code fournisseur I043525G

Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.

Identifiants chimiques

CAS 83-39-6
Molecular Formula C5H6N4O2
Molecular Weight (g/mol) 154.13
MDL Number MFCD00047022
InChI Key NNWAARLSYSBVPB-UHFFFAOYSA-N
Synonym Glycalbylamide
PubChem CID 65745
IUPAC Name 1H-imidazole-4,5-dicarboxamide
SMILES NC(=O)C1=C(N=CN1)C(N)=O

Spécifications

Color White-Yellow
Quantity 25 g
Formula Weight 154.13
Percent Purity ≥97.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Imidazole-4,5-dicarboxamide
missing translation for 'productTitle'
missing translation for 'pleaseSelectIssue'

missing translation for 'byClickingSubmit' missing translation for 'privacyPolicy'.