accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3ONCH3OH3CNNH3C(E/Z)

CAS RN 83-63-6

IUPAC Name: N-acetyl-N-{2-methyl-4-[2-(2-methylphenyl)diazen-1-yl]phenyl}acetamide
Synonyms: N-acetyl-N-{2-methyl-4-[2-(2-methylphenyl)diazen-1-yl]phenyl}acetamide
Molecular Weight (g/mol): 309.37
Molecular Formula: C18H19N3O2
InChi Key: YIEDSISPYKQADU-UHFFFAOYSA-N
SMILES: CC(=O)N(C(C)=O)C1=CC=C(C=C1C)N=NC1=CC=CC=C1C
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results