Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N,N-Diacetyl-o-aminoazotoluene 90.0+%, TCI America™
SDP

Catalog No. D006325G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D006325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D006325G Supplier TCI America Supplier No. D006325G

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 83-63-6
Molecular Formula C18H19N3O2
Molecular Weight (g/mol) 309.37
MDL Number MFCD00026179
InChI Key YIEDSISPYKQADU-UHFFFAOYSA-N
Synonym Pellidol
PubChem CID 6751
IUPAC Name N-acetyl-N-{2-methyl-4-[2-(2-methylphenyl)diazen-1-yl]phenyl}acetamide
SMILES CC(=O)N(C(C)=O)C1=CC=C(C=C1C)N=NC1=CC=CC=C1C

Specifications

Melting Point 76°C
Quantity 25 g
Formula Weight 309.37
Percent Purity ≥90.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material N,N-Diacetyl-o-aminoazotoluene
EINECSNumber (9)-0536
RTECSNumber AB4230000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.