accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 83-73-8

IUPAC Name: 5,7-diiodoquinolin-8-ol
Synonyms: iodoquinol diodoxyquinoleine iodoquinol usp entero-diyod caswell no. 354 diodoquin diiodohydroxyquinolinum diodoxylin di-quinol searle brand of iodoquinol
Molecular Weight (g/mol): 396.95
Molecular Formula: C9H5I2NO
InChi Key: UXZFQZANDVDGMM-UHFFFAOYSA-N
SMILES: OC1=C2N=CC=CC2=C(I)C=C1I
Grade 
  • (5)
Percent Purity 
  • (5)
Physical Form 
  • (5)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (5)
  • (5)
  • (5)
  • (5)
  • (5)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)