accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OO

CAS RN 830-81-9

IUPAC Name: naphthalen-1-yl acetate
Synonyms: 1-acetoxynaphthalene
Molecular Weight (g/mol): 186.21
Molecular Formula: C12H10O2
InChi Key: VGKONPUVOVVNSU-UHFFFAOYSA-N
SMILES: CC(=O)OC1=C2C=CC=CC2=CC=C1
Percent Purity 
  • (1)
  • (3)
Quantity 
  • (2)
  • (1)
  • (1)
Physical Form 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (1)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)