Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Naphthyl acetate, 99%

Catalog No. AAA1748218
Change view
Click to view available options
Quantity:
50 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1748218 50 g
AAA1748230 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1748218 Supplier Thermo Scientific Chemicals Supplier No. A1748218
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 830-81-9 | C12H10O2 | 186.21 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 830-81-9
Molecular Formula C12H10O2
Molecular Weight (g/mol) 186.21
MDL Number MFCD00003922
InChI Key VGKONPUVOVVNSU-UHFFFAOYSA-N
Synonym 1-naphthyl acetate, 1-acetoxynaphthalene, alpha-naphthyl acetate, 1-naphthol, acetate, a-naphthyl acetate, naphthyl acetate, naphthalene, 1-acetoxy, 1-naphthalenol, acetate, acetic acid 1-naphthyl ester, alpha-naphthol acetate
PubChem CID 13247
IUPAC Name naphthalen-1-yl acetate
SMILES CC(=O)OC1=CC=CC2=CC=CC=C21

Specifications

Melting Point 43°C to 46°C
Flash Point >110°C (230°F)
Quantity 50 g
Beilstein 2046403
Sensitivity Moisture sensitive
Formula Weight 186.21
Percent Purity 99%
Chemical Name or Material 1-Naphthyl acetate
Hazard Category Warning
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
EINECSNumber 212-599-8
RTECSNumber QL2975500
TSCA Yes
Recommended Storage Keep cold

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.