accessibility menu, dialog, popup

UserName

Viewing profile as user:
NaNaOSOOSOOO

CAS RN 831-59-4

IUPAC Name: disodium benzene-1,3-disulfonate
Synonyms: disodium benzene-1,3-disulfonate
Molecular Weight (g/mol): 282.19
Molecular Formula: C6H4Na2O6S2
InChi Key: XWPWZOJBTOJEGW-UHFFFAOYSA-L
SMILES: [Na+].[Na+].[O-]S(=O)(=O)C1=CC(=CC=C1)S([O-])(=O)=O
Molecular Weight (g/mol) 
  • (5)
  • (1)
Percent Purity 
  • (2)
  • (1)
  • (3)
Quantity 
  • (1)
  • (1)
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (2)
  • (3)
  • (5)
  • (1)
  • (2)
  • (1)
  • (3)
  • (1)
  • (1)
  • (2)
  • (2)
  • (2)