accessibility menu, dialog, popup

UserName

Viewing profile as user:
OHO

CAS RN 831-82-3

IUPAC Name: 4-phenoxyphenol
Synonyms: 4-phenoxyphenol p-hydroxydiphenyl ether 4-hydroxydiphenyl ether
Molecular Weight (g/mol): 186.21
Molecular Formula: C12H10O2
InChi Key: ZSBDGXGICLIJGD-UHFFFAOYSA-N
SMILES: OC1=CC=C(OC2=CC=CC=C2)C=C1
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (2)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)