Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Phenoxyphenol, 99%, Thermo Scientific™

Catalog No. AAA1848914
Change view
Click to view available options
Quantity:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1848914 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAA1848914 Supplier Thermo Scientific Chemicals Supplier No. A1848914

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
4-Phenoxyphenol is a part of a group of phenolic compounds that have potential apoptotic activity, which can be used in the development of new treatments for cancer therapy.

Solubility
Soluble in water (partly).

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Specifications

CAS 831-82-3
Boiling Point 177°C to 180°C (9mmHg)
Flash Point 170°C (338°F)
Molecular Formula C12H10O2
Quantity 25g
MDL Number MFCD00002331
Beilstein 2047182
Synonym p-phenoxyphenol, 4-hydroxydiphenyl ether, phenol, 4-phenoxy, p-hydroxydiphenyl ether, phenol, p-phenoxy, hydroquinone monophenyl ether, unii-d83r742gjr, 4-phenyloxy phenol, 4-phenoxy phenol, 4-phenoxy-phenol
Solubility Information Soluble in water (partly).
InChI Key ZSBDGXGICLIJGD-UHFFFAOYSA-N
SMILES C1=CC=C(C=C1)OC2=CC=C(C=C2)O
IUPAC Name 4-phenoxyphenol
Molecular Weight (g/mol) 186.21
PubChem CID 13254
ChEBI CHEBI:39264
Formula Weight 186.21
Percent Purity 99%
Chemical Name or Material 4-Phenoxyphenol
Show More Show Less
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 212-611-1
TSCA Yes
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.