accessibility menu, dialog, popup

UserName

Viewing profile as user:
S

CAS RN 831-91-4

IUPAC Name: (benzylsulfanyl)benzene
Synonyms: benzyl phenyl sulfide
Molecular Weight (g/mol): 200.3
Molecular Formula: C13H12S
InChi Key: LKMCJXXOBRCATQ-UHFFFAOYSA-N
SMILES: C(SC1=CC=CC=C1)C1=CC=CC=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results