Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Benzyl Phenyl Sulfide 98.0+%, TCI America™
SDP

Catalog No. B012710G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B012710G 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B012710G Supplier TCI America Supplier No. B012710G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 831-91-4
Molecular Formula C13H12S
Molecular Weight (g/mol) 200.30
MDL Number MFCD00003066
InChI Key LKMCJXXOBRCATQ-UHFFFAOYSA-N
Synonym benzyl phenyl sulfide, benzyl phenyl sulfane, phenyl benzyl sulfide, benzylphenylsulfide, benzyl phenyl sulphide, benzylsulfanyl benzene, sulfide, benzyl phenyl, benzene, phenylmethyl thio, benzylphenyl sulfide, phenylmethylthio benzene
PubChem CID 13255
IUPAC Name (benzylsulfanyl)benzene
SMILES C(SC1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 43°C
Color White-Yellow
Boiling Point 197°C
Quantity 10 g
Formula Weight 200.30
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Benzyl Phenyl Sulfide
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.