accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOCH3CH3OOH3C

CAS RN 832-58-6

IUPAC Name: 1-(2,4,6-trimethoxyphenyl)ethan-1-one
Synonyms: 1-(2,4,6-trimethoxyphenyl)ethanone
Molecular Weight (g/mol): 210.23
Molecular Formula: C11H14O4
InChi Key: KPZWHZSIXZXDMW-UHFFFAOYSA-N
SMILES: COC1=CC(OC)=C(C(C)=O)C(OC)=C1
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)