Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,4,6-Trimethoxyacetophenone, MP Biomedicals

Catalog No. ICN225761 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN225761 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN225761 Supplier MP Biomedicals, Inc Supplier No. 0522576105

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 832-58-6
Molecular Formula C11H14O4
Molecular Weight (g/mol) 210.23
MDL Number MFCD00017238
InChI Key KPZWHZSIXZXDMW-UHFFFAOYSA-N
Synonym 2',4',6'-trimethoxyacetophenone, 1-2,4,6-trimethoxyphenyl ethanone, 2,4,6-trimethoxyacetophenone, ethanone, 1-2,4,6-trimethoxyphenyl, ethanone,1-2,4,6-trimethoxyphenyl, 1-acetyl-2,4,6-trimethoxybenzene, o-methylxanthoxylin, cambridge id 5135317, 2,4,6-trimethoxy acetophenone, 2/',4/',6/'-trimethoxyacetophenone
PubChem CID 123089
IUPAC Name 1-(2,4,6-trimethoxyphenyl)ethan-1-one
SMILES COC1=CC(OC)=C(C(C)=O)C(OC)=C1

Specifications

Quantity 10 g
Formula Weight 210.23
Chemical Name or Material 2, 4, 6-Trimethoxy Acetophenone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.