accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 832-71-3

IUPAC Name: 3-methylphenanthrene
Synonyms: 3-methylphenanthrene
Molecular Weight (g/mol): 192.26
Molecular Formula: C15H12
InChi Key: GKYWZUBZZBHZKU-UHFFFAOYSA-N
SMILES: CC1=CC=C2C=CC3=CC=CC=C3C2=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results