Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Methylphenanthrene 98.0+%, TCI America™
SDP

Catalog No. M0523100MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M0523100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M0523100MG Supplier TCI America Supplier No. M0523100MG
Add to Cart
Add to Cart

Chemical Identifiers

CAS 832-71-3
Molecular Formula C15H12
Molecular Weight (g/mol) 192.261
MDL Number MFCD00216221
InChI Key GKYWZUBZZBHZKU-UHFFFAOYSA-N
PubChem CID 13258
IUPAC Name 3-methylphenanthrene
SMILES CC1=CC2=C(C=C1)C=CC3=CC=CC=C32

Specifications

Melting Point 63°C
Color White
Boiling Point 370°C
Quantity 100 mg
Formula Weight 192.26
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3-Methylphenanthrene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.