accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OOCH3

CAS RN 83242-83-5

IUPAC Name: 4-propylcyclohexyl 4'-propyl-[1,1'-bi(cyclohexane)]-4-carboxylate
Synonyms: 4-propylcyclohexyl 4'-propyl-[1,1'-bi(cyclohexane)]-4-carboxylate
Molecular Weight (g/mol): 376.63
Molecular Formula: C25H44O2
InChi Key: ITVBZSWXRMMPFS-UHFFFAOYSA-N
SMILES: CCCC1CCC(CC1)OC(=O)C1CCC(CC1)C1CCC(CCC)CC1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results