Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

trans-4-Propylcyclohexyl trans,trans-4'-Propylbicyclohexyl-4-carboxylate 98.0+%, TCI America™
SDP

Catalog No. P24365G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P24365G 5 g
P24361G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P24365G Supplier TCI America Supplier No. P24365G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 83242-83-5
Molecular Formula C25H44O2
Molecular Weight (g/mol) 376.625
MDL Number MFCD20527218
InChI Key ITVBZSWXRMMPFS-UHFFFAOYSA-N
Synonym 4-propylcyclohexyl 4-4-propylcyclohexyl cyclohexane-1-carboxylate, 4-propylcyclohexyl trans trans trans-4'-propyl 1,1'-bicyclohexyl-4-carboxylate, trans-4-propylcyclohexyl trans,trans-4'-propylbicyclohexyl-4-carboxylate, trans,trans-trans-4-propylcyclohexyl 4'-propyl-1,1'-bi cyclohexane-4-carboxylate, 4-propylcyclohexyl trans trans trans-4-propyl 1,1-bicyclohexyl-4-carboxylate, trans,trans-4'-propylbicyclohexyl-4-carboxylic acid trans-4-propylcyclohexyl ester, trans-4-n-propylcyclohexyl trans,trans-4'-n-propylbicyclohexyl-4-carboxylate, 1s,4r-4-propylcyclohexyl 1r,1's,4r,4'r-4'-propyl-1,1'-bi cyclohexane-4-carboxylate, 1,1'-bicyclohexyl-4-carboxylic acid, 4'-propyl-, 4-propylcyclohexyl ester, trans trans trans, 1,1'-bicyclohexyl-4-carboxylicacid, 4'-propyl-, trans-4-propylcyclohexyl ester, trans,trans
PubChem CID 550127
IUPAC Name (4-propylcyclohexyl) 4-(4-propylcyclohexyl)cyclohexane-1-carboxylate
SMILES CCCC1CCC(CC1)C2CCC(CC2)C(=O)OC3CCC(CC3)CCC

Specifications

Melting Point 170°C
Color White
Quantity 5 g
Formula Weight 376.63
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material trans-4-Propylcyclohexyl trans,trans-4′-Propylbicyclohexyl-4-carboxylate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.