accessibility menu, dialog, popup

UserName

Viewing profile as user:
FFFFFFFFFHO

CAS RN 83310-97-8

IUPAC Name: 4,4,5,5,6,6,7,7,7-nonafluoroheptan-1-ol
Synonyms: 4,4,5,5,6,6,7,7,7-nonafluoroheptan-1-ol
Molecular Weight (g/mol): 278.12
Molecular Formula: C7H7F9O
InChi Key: OVBNEUIFHDEQHD-UHFFFAOYSA-N
SMILES: OCCCC(F)(F)C(F)(F)C(F)(F)C(F)(F)F
Boiling Point 
  • (1)
  • (3)
Molecular Weight (g/mol) 
  • (3)
  • (1)
Quantity 
  • (1)
  • (2)
  • (1)
Percent Purity 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (1)
  • (3)
  • (3)
  • (1)
  • (1)
  • (2)
  • (1)
  • (3)
  • (3)