CAS RN 83380-82-9
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
MilliporeSigma™ Calbiochem™ Vitronectin, Human Plasma
Intact protein and SDS-PAGE bands react with monoclonal antibodies to vitronectin, bind to heparin, and promote cell adhesion