Learn More
Description
- Native vitronectin purified from human plasma
- Extracellular matrix protein that promotes cell adhesion
- Exists as a mixture of 75 kDa and 65 kDa polypeptides
- Biochemically and immunologically distinct from fibronectin
- Binds to cells through the RGD sequence in its cell-binding domain
Specifications
Specifications
| CAS | 83380-82-9 |
| Content And Storage | Store at −20°C |
| Purity | ≥95% by SDS-PAGE |
| Quantity | 100 μg |
| Form | Lyophilized |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.