CAS RN 83480-29-9
IUPAC Name:
(1S,2S,3R,4S,5S)-5-[(1,3-dihydroxypropan-2-yl)amino]-1-(hydroxymethyl)cyclohexane-1,2,3,4-tetrol
Synonyms:
voglibose voglibosa vocarb basen voglibosum volix basen (takeda chemical industries) volix (ranbaxy labs) 伏格列波糖 glustat
Molecular Weight (g/mol):
267.28
Molecular Formula:
C10H21NO7
InChi Key:
FZNCGRZWXLXZSZ-CIQUZCHMSA-N
SMILES:
OCC(CO)N[C@H]1C[C@](O)(CO)[C@@H](O)[C@H](O)[C@H]1O
1
–
1
of
1
results
1
Voglibose 98.0+%, TCI America™
CAS: 83480-29-9 Molecular Formula: C10H21NO7 Molecular Weight (g/mol): 267.278 MDL Number: MFCD00865496 InChI Key: FZNCGRZWXLXZSZ-CIQUZCHMSA-N PubChem CID: 444020 IUPAC Name: (1S,2S,3R,4S,5S)-5-(1,3-dihydroxypropan-2-ylamino)-1-(hydroxymethyl)cyclohexane-1,2,3,4-tetrol SMILES: C1C(C(C(C(C1(CO)O)O)O)O)NC(CO)CO