Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Voglibose 98.0+%, TCI America™
SDP

Catalog No. V0119100MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
V0119100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. V0119100MG Supplier TCI America Supplier No. V0119100MG
Add to Cart
Add to Cart

Chemical Identifiers

CAS 83480-29-9
Molecular Formula C10H21NO7
Molecular Weight (g/mol) 267.278
MDL Number MFCD00865496
InChI Key FZNCGRZWXLXZSZ-CIQUZCHMSA-N
PubChem CID 444020
IUPAC Name (1S,2S,3R,4S,5S)-5-(1,3-dihydroxypropan-2-ylamino)-1-(hydroxymethyl)cyclohexane-1,2,3,4-tetrol
SMILES C1C(C(C(C(C1(CO)O)O)O)O)NC(CO)CO

Specifications

Melting Point 165°C
Color White
Quantity 100 mg
Formula Weight 267.28
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Voglibose
RTECSNumber NM7524600
TSCA No
Recommended Storage Freezer
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.