accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 835-64-3

IUPAC Name: 6-[(2E)-2,3-dihydro-1,3-benzoxazol-2-ylidene]cyclohexa-2,4-dien-1-one
Synonyms: 6-[(2E)-3H-1,3-benzoxazol-2-ylidene]cyclohexa-2,4-dien-1-one
Molecular Weight (g/mol): 211.22
Molecular Formula: C13H9NO2
InChi Key: UWUSYRFTCSSHAJ-UKTHLTGXSA-N
SMILES: O=C1C=CC=C\C1=C1\NC2=CC=CC=C2O1
Quantity 
  • (1)
  • (1)
  • (2)
Boiling Point 
  • (2)
  • (1)
Percent Purity 
  • (2)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)