Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-(o-Hydroxyphenyl)benzoxazole,MP Biomedicals

Catalog No. ICN204715 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN204715 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN204715 Supplier MP Biomedicals, Inc Supplier No. 0520471505

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 835-64-3
Molecular Formula C13H9NO2
Molecular Weight (g/mol) 211.22
InChI Key UWUSYRFTCSSHAJ-UHFFFAOYSA-N
Synonym 2-2-hydroxyphenyl benzoxazole, 6-3h-1,3-benzoxazol-2-ylidene cyclohexa-2,4-dien-1-one, 2-2-benzoxazolyl phenol, ghgzvwotjdlrey-uhfffaoysa-n, phenol, benzoxazolyl, acmc-209psp, 2-2'-hydroxyphenyl benzoxazole, 2,4-cyclohexadien-1-one, 6-2 3h-benzoxazolylidene
PubChem CID 6744797
IUPAC Name 6-(3H-1,3-benzoxazol-2-ylidene)cyclohexa-2,4-dien-1-one
SMILES C1=CC=C2C(=C1)NC(=C3C=CC=CC3=O)O2

Specifications

Quantity 10 g
Formula Weight 211.22
Chemical Name or Material 2-(O-Hydroxyphenyl)Benzoxazole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.