accessibility menu, dialog, popup

UserName

Viewing profile as user:
OOHHOOH(E/Z)

CAS RN 83616-07-3

IUPAC Name: 3-(2,4-dihydroxyphenyl)-1-(4-hydroxyphenyl)prop-2-en-1-one
Synonyms: 3-(2,4-dihydroxyphenyl)-1-(4-hydroxyphenyl)prop-2-en-1-one
Molecular Weight (g/mol): 256.26
Molecular Formula: C15H12O4
InChi Key: VDYSHUXENHRSOO-UHFFFAOYSA-N
SMILES: OC1=CC=C(C=C1)C(=O)C=CC1=C(O)C=C(O)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results