Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,4,4'-Trihydroxybenzalacetophenone, MP Biomedicals

Catalog No. ICN221722 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN221722 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN221722 Supplier MP Biomedicals, Inc Supplier No. 0522172210

Chemical Identifiers

CAS 83616-07-3
Molecular Formula C15H12O4
Molecular Weight (g/mol) 256.26
MDL Number MFCD00049007
InChI Key VDYSHUXENHRSOO-UHFFFAOYSA-N
Synonym 2,4,4'-trihydroxychalcone, 3-2,4-dihydroxyphenyl-1-4-hydroxyphenyl prop-2-en-1-one, 2,4,4'-trihydroxy benzalacetophenone, 2-propen-1-one, 3-2,4-dihydroxyphenyl-1-4-hydroxyphenyl
PubChem CID 5057077
IUPAC Name 3-(2,4-dihydroxyphenyl)-1-(4-hydroxyphenyl)prop-2-en-1-one
SMILES OC1=CC=C(C=C1)C(=O)C=CC1=C(O)C=C(O)C=C1

Specifications

Quantity 10 g
Chemical Name or Material 2, 4, 4«-Trihydroxy Benzalacetophenone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.