accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 838-07-3

IUPAC Name: 4-amino-1-[(2R,4S,5R)-4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]-5-methyl-1,2-dihydropyrimidin-2-one
Synonyms: 5-methyldeoxycytidine
Molecular Weight (g/mol): 241.25
Molecular Formula: C10H15N3O4
InChi Key: LUCHPKXVUGJYGU-XLPZGREQSA-N
SMILES: CC1=CN([C@H]2C[C@H](O)[C@@H](CO)O2)C(=O)N=C1N
Molecular Weight (g/mol) 
  • (2)
  • (4)
  • (1)
Physical Form 
  • (3)
  • (4)
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (3)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (1)
  • (2)
  • (4)
  • (1)
  • (3)
  • (4)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (3)
  • (1)