Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Methyl-2'-deoxycytidine, MP Biomedicals™

Catalog No. MP219888380 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
10 mg
25 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP219888380 100 mg
ICN19888310 10 mg
ICN19888325 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. MP219888380 Supplier MP Biomedicals, Inc Supplier No. 0219888380
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

5-Methyl-2'-deoxycytidine, White Powder

  • 5-Methyl-2'-deoxycytidine in single-stranded DNA can act in cis to signal de novo DNA methylation.
  • It has been used in epigenetics research to investigate the dynamics of DNA methylation pattern in the control of gene expression.
  • A cytidine analog and an isostere of thymidine.
  • 5-Methyl-2'-deoxycytidine is a pyrimidine nucleoside that when incorporated into single-stranded DNA can act in cis to signal de novo DNA methylation.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 838-07-3
MDL Number MFCD00006549
InChI Key LUCHPKXVUGJYGU-XLPZGREQSA-N

Specifications

Melting Point 217°C to 219°C (literature)
Color White
Molecular Formula C10H15N3O4
Refractive Index 1.694 (lit.)
Quantity 100 mg
Synonym 2'-deoxy-5-methyl-cytidine
Solubility Information Soluble in 1 N NaOH.
Optical Rotation [α]25/D +63°, c = 1 in 1 M NaOH (lit.)
Molecular Weight (g/mol) 241.25
Vapor Pressure 8.85E-12mmHg at 25°C (lit.)
Physical Form Powder
Grade 99.6 area% (HPLC)
Show More Show Less
RTECSNumber HA3860000
storageNote1 Store at Room Temperature (15-30°C).
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.