accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CSH3C

CAS RN 838-59-5

IUPAC Name: {1-[(1-phenylethyl)sulfanyl]ethyl}benzene
Synonyms: {1-[(1-phenylethyl)sulfanyl]ethyl}benzene
Molecular Weight (g/mol): 242.38
Molecular Formula: C16H18S
InChi Key: NTKXWHALINJHQX-UHFFFAOYNA-N
SMILES: CC(SC(C)C1=CC=CC=C1)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results