Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Di(alpha-phenylethyl) Sulfide (DL- and meso- mixture), TCI America™
SDP

Catalog No. D08895G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D08895G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D08895G Supplier TCI America Supplier No. D08895G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 838-59-5
Molecular Formula C16H18S
Molecular Weight (g/mol) 242.38
MDL Number MFCD00026342
InChI Key NTKXWHALINJHQX-UHFFFAOYSA-N
Synonym 1-Phenylethyl Sulfide
PubChem CID 248893
IUPAC Name 1-(1-phenylethylsulfanyl)ethylbenzene
SMILES CC(C1=CC=CC=C1)SC(C)C2=CC=CC=C2

Specifications

Quantity 5 g
Formula Weight 242.38
Physical Form Liquid
Chemical Name or Material Di(alpha-phenylethyl) Sulfide (DL- and meso- mixture)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.