accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 838-88-0

IUPAC Name: 4-[(4-amino-3-methylphenyl)methyl]-2-methylaniline
Synonyms: 4,4'-methylenedi-o-toluidine
Molecular Weight (g/mol): 226.32
Molecular Formula: C15H18N2
InChi Key: WECDUOXQLAIPQW-UHFFFAOYSA-N
SMILES: CC1=CC(CC2=CC=C(N)C(C)=C2)=CC=C1N
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results