Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4,4'-Diamino-3,3'-dimethyldiphenylmethane 97.0+%, TCI America™
SDP

Catalog No. D204625G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D204625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D204625G Supplier TCI America Supplier No. D204625G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 838-88-0
Molecular Formula C15H18N2
Molecular Weight (g/mol) 226.323
MDL Number MFCD00126963
InChI Key WECDUOXQLAIPQW-UHFFFAOYSA-N
Synonym 4,4′C-Methylenebis-o-toluidine
PubChem CID 13283
ChEBI CHEBI:82343
IUPAC Name 4-[(4-amino-3-methylphenyl)methyl]-2-methylaniline
SMILES CC1=C(C=CC(=C1)CC2=CC(=C(C=C2)N)C)N

Specifications

Melting Point 156°C
Quantity 25 g
UN Number 3077
Formula Weight 226.32
Percent Purity ≥97.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 4,4′-Diamino-3,3′-dimethyldiphenylmethane
EINECSNumber (4)-0863
RTECSNumber BY5300000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.