accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 838-96-0

IUPAC Name: 2-[4-(benzyloxy)phenyl]acetonitrile
Synonyms: 2-[4-(benzyloxy)phenyl]acetonitrile
Molecular Weight (g/mol): 223.28
Molecular Formula: C15H13NO
InChi Key: QKEYZRVDFZDOEP-UHFFFAOYSA-N
SMILES: N#CCC1=CC=C(OCC2=CC=CC=C2)C=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results