accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3NNNHN(E/Z)(E/Z)

CAS RN 83809-73-8

IUPAC Name: N-[(2-methylphenyl)imino]-N'-(phenylamino)benzenecarboximidamide
Synonyms: N-[(2-methylphenyl)imino]-N'-(phenylamino)benzenecarboximidamide
Molecular Weight (g/mol): 314.39
Molecular Formula: C20H18N4
InChi Key: KSUKLVGDPSNHPH-UHFFFAOYSA-N
SMILES: CC1=CC=CC=C1N=NC(=NNC1=CC=CC=C1)C1=CC=CC=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results