accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3IH2N

CAS RN 83863-33-6

IUPAC Name: 5-iodo-2-methylaniline
Synonyms: 5-iodo-2-methylaniline
Molecular Weight (g/mol): 233.05
Molecular Formula: C7H8IN
InChi Key: IOEHXNCBPIBDBZ-UHFFFAOYSA-N
SMILES: CC1=CC=C(I)C=C1N

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results