Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Iodo-2-methylaniline, 97%, Thermo Scientific™

Catalog No. AC442430010
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC442430010 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC442430010 Supplier Thermo Scientific Chemicals Supplier No. 442430010

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 83863-33-6
Molecular Formula C7H8IN
Molecular Weight (g/mol) 233.05
MDL Number MFCD00051531
InChI Key IOEHXNCBPIBDBZ-UHFFFAOYSA-N
Synonym 2-amino-4-iodotoluene, 5-iodo-o-toluidine, 5-iodo-2-methyl-phenylamine, benzenamine, 5-iodo-2-methyl, benzenamine,5-iodo-2-methyl, 5-iodo-2-methyl-aniline, 5-iodo-2-methylbenzenamine, 5-iodo-2-methylphenylamine, 2-methyl-5-iodoaniline, #
PubChem CID 522802
IUPAC Name 5-iodo-2-methylaniline
SMILES CC1=CC=C(I)C=C1N

Specifications

Melting Point 48°C to 50°C
Flash Point >110°C
Assay Percent Range 96% min. (GC)
Packaging Glass Bottle
Quantity 1 g
Formula Weight 233.05
Percent Purity 97%
Chemical Name or Material 5-Iodo-2-methylaniline
Hazard Category GHS Signal Word: Danger
EINECSNumber 281-094-2

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.