accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CBOCH3CH3CH3H3CO(E)

CAS RN 83947-55-1

IUPAC Name: 4,4,5,5-tetramethyl-2-[(1E)-oct-1-en-1-yl]-1,3,2-dioxaborolane
Synonyms: 4,4,5,5-tetramethyl-2-[(1E)-oct-1-en-1-yl]-1,3,2-dioxaborolane
Molecular Weight (g/mol): 238.18
Molecular Formula: C14H27BO2
InChi Key: KQTOSGTXAFJZSJ-VAWYXSNFSA-N
SMILES: CCCCCC\C=C\B1OC(C)(C)C(C)(C)O1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results