accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 84-15-1

IUPAC Name: 2-phenyl-1,1'-biphenyl
Synonyms: O-terphenyl
Molecular Weight (g/mol): 230.31
Molecular Formula: C18H14
InChi Key: OIAQMFOKAXHPNH-UHFFFAOYSA-N
SMILES: C1=CC=C(C=C1)C1=CC=CC=C1C1=CC=CC=C1
Molecular Weight (g/mol) 
  • (2)
  • (6)
Boiling Point 
  • (3)
  • (2)
Percent Purity 
  • (3)
  • (2)
Quantity 
  • (2)
  • (2)
  • (1)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (3)
  • (2)
  • (6)
  • (3)
  • (2)
  • (3)
  • (2)
  • (2)
  • (2)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)