Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

o-Terphenyl 99.0+%, TCI America™
SDP

Catalog No. T0019100G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T0019100G 100 g
T001925G 25 g
T0019500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. T0019100G Supplier TCI America Supplier No. T0019100G
Only null left
Add to Cart
Add to Cart

Chemical Identifiers

CAS 84-15-1
Molecular Formula C18H14
Molecular Weight (g/mol) 230.31
MDL Number MFCD00003055
InChI Key OIAQMFOKAXHPNH-UHFFFAOYSA-N
Synonym o-terphenyl, 1,1':2',1-terphenyl, terphenyl, chlorinated, 1,1'-biphenyl, 2-phenyl, caswell no. 672b, o-diphenylbenzene, polychlorinated terphenyl, polychlorinated terphenyls, unii-w5675r7kvw, ccris 1655
PubChem CID 6766
IUPAC Name 1,2-diphenylbenzene
SMILES C1=CC=C(C=C1)C2=CC=CC=C2C3=CC=CC=C3

Specifications

Melting Point 58°C
Color White
Boiling Point 332°C
Quantity 100 g
UN Number 3077
Formula Weight 230.31
Percent Purity ≥99.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material o-Terphenyl
EINECSNumber (4)-0017
RTECSNumber WZ6472000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.