accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 84102-69-2

IUPAC Name: ethyl 5-bromo-1-benzofuran-2-carboxylate
Synonyms: ethyl 5-bromo-1-benzofuran-2-carboxylate
Molecular Weight (g/mol): 269.09
Molecular Formula: C11H9BrO3
InChi Key: XLJWAHXKBCDQNP-UHFFFAOYSA-N
SMILES: CCOC(=O)C1=CC2=CC(Br)=CC=C2O1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results