Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl(5-bromobenzofuran)-2-carboxylate, 95%, Thermo Scientific™

Catalog No. CC31223DA
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CC31223DA 1 g
CC31223CB 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. CC31223DA Supplier Thermo Scientific Chemicals Supplier No. CC31223DA
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 84102-69-2
Molecular Formula C11H9BrO3
Molecular Weight (g/mol) 269.094
InChI Key XLJWAHXKBCDQNP-UHFFFAOYSA-N
PubChem CID 735184
IUPAC Name ethyl 5-bromo-1-benzofuran-2-carboxylate
SMILES CCOC(=O)C1=CC2=C(O1)C=CC(=C2)Br

Specifications

Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 1 g
Synonym ethyl 5-bromobenzofuran-2-carboxylate, buttpark 34\07-75, 5-bromo-benzofuran-2-carboxylic acid ethyl ester, ethylbromobenzofurancarboxylate, ethyl5-bromobenzofuran-2-carboxylate, ethyl 5-bromo-2-benzofurancarboxylate, ethyl 5-bromo-benzofuran-2-carboxylate, 5-bromo-2-ethoxycarbonyl-1-benzofuran
Formula Weight 269.09
Percent Purity 95%
Chemical Name or Material Ethyl (5-bromobenzofuran)-2-carboxylate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.