accessibility menu, dialog, popup

UserName

Viewing profile as user:
FOCl

CAS RN 84194-30-9

IUPAC Name: 2-chloro-5-fluorobenzaldehyde
Synonyms: 2-chloro-5-fluorobenzaldehyde
Molecular Weight (g/mol): 158.56
Molecular Formula: C7H4ClFO
InChi Key: SOEFVBXUNROUOX-UHFFFAOYSA-N
SMILES: FC1=CC(C=O)=C(Cl)C=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results