Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Chloro-5-fluorobenzaldehyde 98.0+%, TCI America™
SDP

Catalog No. C23815G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C23815G 5 g
C238125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C23815G Supplier TCI America Supplier No. C23815G

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 84194-30-9
Molecular Formula C7H4ClFO
Molecular Weight (g/mol) 158.56
MDL Number MFCD03788511
InChI Key SOEFVBXUNROUOX-UHFFFAOYSA-N
PubChem CID 2779184
IUPAC Name 2-chloro-5-fluorobenzaldehyde
SMILES FC1=CC(C=O)=C(Cl)C=C1

Specifications

Melting Point 48°C
Color White-Yellow
Quantity 5 g
Formula Weight 158.56
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Chloro-5-fluorobenzaldehyde
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.